missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAPTM4B Antibody (CL5289) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61158UV
This item is not returnable.
View return policy
Description
LAPTM4B Monoclonal antibody specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| LAPTM4B | |
| Monoclonal | |
| DyLight 350 | |
| LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta | |
| This antibody was developed using a synthetic peptide derived from Q86VI4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 55353 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5289 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction