missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP (TGF-beta 1) Antibody (7F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
245.00€ - 571.00€
Specifications
| Antigen | LAP (TGF-beta 1) |
|---|---|
| Clone | 7F6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry reported in scientific literature (PMID 30147399), Immunohistochemistry-Paraffin reported by customer review |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18779963
|
Novus Biologicals
NBP2-22114SS |
0.025 mL |
245.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757313
|
Novus Biologicals
NBP2-22114 |
0.1 mL |
571.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LAP (TGF-beta 1) | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology | |
| PBS with 0.02% Sodium Azide | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| TGFB1 | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 44.3 kDa |
| 7F6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry reported in scientific literature (PMID 30147399), Immunohistochemistry-Paraffin reported by customer review | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human, Mouse, Rat | |
| P01137 | |
| 7040 | |
| Purified recombinant fragment of human TGF beta 1 (amino acids 62-197) expressed in E. coli. [UniProt# P01137] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title