missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (CL4489) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59053PCP
This item is not returnable.
View return policy
Description
LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| LAMP-1/CD107a | |
| Monoclonal | |
| PerCP | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| This antibody was developed using a synthetic peptide derived from P11279, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| 3916 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4489 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction