Learn More
Laminin alpha 5 Antibody (CL3118) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42391JF669
Product Code. 30299150
  • 770.00€ / 0.10mL
Description
Laminin alpha 5 Monoclonal antibody specifically detects Laminin alpha 5 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha 5
Monoclonal
Janelia Fluor 669
KIAA0533, KIAA1907, laminin alpha-5 chain, laminin subunit alpha-5, laminin, alpha 5, Laminin-10 subunit alpha, Laminin-11 subunit alpha, Laminin-15 subunit alpha
This antibody was developed using a recombinant protein derived from O15230, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Rat
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL3118
50mM Sodium Borate
Mouse
Protein A purified
RUO
3911
Store at 4⪚C in the dark.