missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C Antibody (mab636), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-74451-50ul
165.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Lamin A + C Monoclonal specifically detects Lamin A + C in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Lamin A + C | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| Mouse | |
| Unpurified | |
| RUO | |
| 4000 | |
| Human | |
| Supernatant |
| Western Blot | |
| mab636 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Frozen 1:100 | |
| Q3ZD69 | |
| LMNA | |
| Porcine lamin preparation. | |
| 50 μL | |
| Primary | |
| Detects lamin A/C from human, bovine and porcine samples. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction