missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (IGL/9397R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24091PCP
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| PerCP | |
| IGLC, immunoglobulin lambda constant group | |
| Recombinant fragment (around aa1-200) of human Lambda Light Chain protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| IGL/9397R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction