missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lactase Antibody (CL14133), Novus Biologicals™

Mouse Monoclonal Antibody

387.00€ - 582.00€

Specifications

Antigen Lactase
Clone CL14133
Concentration 1 mg/mL
Dilution Immunohistochemistry 1:2500 - 1:5000
Applications Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043267
View Documents
Novus Biologicals™
NBP3-26844-100ul
100 μL
582.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043846
View Documents
Novus Biologicals™
NBP3-26844-25ul
25 μL
387.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

Lactase
1 mg/mL
Immunohistochemistry
Unconjugated
Mouse
Cancer, Cellular Signaling, metabolism
PBS, pH 7.2, 40% glycerol
3938
IgG2a
Protein A purified
CL14133
Immunohistochemistry 1:2500 - 1:5000
Monoclonal
Purified
RUO
Human
EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1
This antibody was developed against a Synthetic Peptide of P09848. Actual immunogen sequence is proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.