missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | Lactase |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL14122 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P09848, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?