Learn More
Labconco™ Protector XVS™ Flexible Duct Connection for Ventilation Stations
Product Code. 15429484 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector VS Ventilation Station
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15429484 Protector VS Ventilation Station Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15429484 Supplier Labconco™ Supplier No. 4861800
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Flexible Duct Connection for Ventilation Stations
  • For use with Labconco Protector XVS Ventilation Stations
  • Flexible duct connection reduces vibration between blower and PVC ductwork; supplied with two installation clamps
Specifications
For Use With (Equipment) Protector VS Ventilation Station
Height (English) 12 in.
Weight (English) 8 lb.
Weight (Metric) 3.6 Kg
Height (Metric) 30.48 cm
Includes Flexible Duct Connection
Dimensions 6 in. dia. round PVC duct
Material PVC
For Use With (Application) Ducting