Learn More
Labconco™ Flat Work Surface
Product Code. 16647142 Shop All Labconco Corporation Products
Change view
Click to view available options
Width (English):
72 in.
Weight (English):
205 lb.
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Weight (English) missing translation for 'unitSize'
16647142 72 in. 205 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 16647142 missing translation for 'mfr' Labconco™ missing translation for 'supplierNo' 4882804
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Extend your work space with the Labconco Flat Work Surface, perfect when paired with the Labconco Fiberglass 30 and Basic™ hoods
Extend your work space with the Labconco Flat Work Surface, perfect when paired with the Labconco Fiberglass 30 and Basic™ hoods
Install atop Labconco Protector™ storage cabinets and adjustable height base stands.
  • Placed atop two Protector standard base cabinets to support precise glove boxes
  • Flat, solid-epoxy work surfaces
Specifications
For Use With (Equipment) Basic 70 laboratory hood, Precise™ and XPert™ weigh box
Height (English) 1.25 in.
Weight (English) 205 lb.
Weight (Metric) 93 kg
Length (English) 30 in.
Depth (English) 30 in.
Width (English) 72 in.
Height (Metric) 3 cm
Depth (Metric) 76 cm
Width (Metric) 183 cm
Product Type Flat Work Surface
Material Epoxy
Color Black
Cut Style Without Cutout