missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D Receptor (VDR) Ab-1 Rat Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Vitamin D Receptor (VDR) Ab-1, Rat Monoclonal Antibody.

Brand:  Epredia RT-200-B

 View more versions of this product

Product Code. 12633847

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

VDR is a member of super family of steroid hormone receptors. It is known to play role in cellular growth and differentiation, calcium and phosphorus metabolism and bone remodeling. Human breast carcinomas, leukemias, and prostatic carcinoma cells express VDR, which can modify their growth and differentiation. In avians, VDR appears as a doublet of 58kDa and 60kDa.

Host Species: Rat

Clone: 9A7γ.E10.E4

Isotype: IgG2b

Species Reactivity: Human, Pig, Mouse, Rat, Hamster, Frog, Chicken and Fish. Others not known.

Epitope: aa 89-105 (near the DNA-binding domain)

Immunogen: Vitamin D receptor protein from chicken intestine

Molecular Weight: 53kDa

Positive Control: LNCAP cells, HeLa cells

Cellular Localization: Nuclear Ab-1 reacts with both the occupied and unoccupied forms of VDR and shows no cross reaction with estrogen and glucocorticoid receptors. It is excellent for biochemical and histochemical applications.

Recommended for:

  • Western Blotting
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vitamin D Receptor (VDR) Ab-1
Monoclonal
200µg/mL
Rat
1 mL
Nuclear Receptors
Chicken, Fish, Frog, Hamster, Human, Mouse, Pig
IgG2b
Western Blot
9A7gamma.E10.E4
Biotin
Vitamin D receptor protein from chicken intestine
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.