Learn More
Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli
Molecular Weight: 10-12kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic, membrane and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | S100 A4 Ab-8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Purified recombinant human S100 A4 monomer expressed in E. coli |
| Purification Method | Tonsil |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.