missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide

Product Code. 12673147 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
100µL
1mL
500µL
7mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12673147 100 μL Concentrated 100µL
12683147 500 μL Concentrated 500µL
12663147 1 mL Concentrated 1mL
12613157 7 mL Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12673147 Supplier Epredia™ Supplier No. RB1804A0

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Epredia™ S100 A4 Ab-8, Rabbit Polyclonal Antibody provides accurate, reproducible results.

The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic, membrane and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen S100 A4 Ab-8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified recombinant human S100 A4 monomer expressed in E. coli
Purification Method Tonsil
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Equine, Feline, Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.