missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Ab-6 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.

Brand:  Epredia MS-196-PABX

 View more versions of this product

Product Code. 12650497

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 6

Isotype: IgG2b

Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: Not determined

Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells

Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Progesterone Receptor Ab-6
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Bovine, Chicken, Human, Rabbit
IgG2b
Immunofluorescence, Immunoprecipitation
hPRa 6
Unconjugated
Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.