Learn More
Progesterone Receptor Ab-6 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.
Brand: Epredia MS-196-P1ABX
Description
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.Host Species: Mouse
Clone: hPRa 6
Isotype: IgG2b
Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.
Epitope: Not determined
Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (triplet, B-form)
Positive Control: T47D cells
Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.
Recommended for:
- Western Blotting (Not optimum)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Note:
USA: RUO; Int'l: RUO
Specifications
| Progesterone Receptor Ab-6 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Bovine, Chicken, Human, Rabbit | |
| IgG2b |
| Immunofluorescence, Immunoprecipitation | |
| hPRa 6 | |
| Unconjugated | |
| Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.