missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ NY-BR-1 Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Epredia™ NY-BR-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1932-P0

 View more versions of this product

Product Code. 12610487

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

NY-BR-1 is a novel differentiation marker predominantly expressed in normal breast and breast cancer. It has been shown by Immunohistochemistry (IHC) that NY-BR-1 was present solely in ductal epithelium of normal breast tissue. Invasive carcinoma of the breast and carcinoma in situ were positive for NY-BR-1, whereas most other tumors and normal tissues are negative, with the exceptions of one-third of sweat gland carcinomas and around two percent of prostate carcinoma. NY-BR-1 is reported to express more frequently in estrogen-positive and lymph node-negative primary carcinomas, and more commonly in grade 1 than in grade 2 or grade 3. In a study by Varga et al (2006) forty-nine percent of lymph node metastasis were shown to be positive.

Host Species: Mouse

Clone: NY-BR-1No. 2

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: a 77 aa peptide from human NYBR-1 protein

Molecular Weight: 150-160kDa

Positive Control: Breast carcinoma

Cellular Localization: Predominantly cytoplasmic and occasional nuclear

Recommended for:

Immunohistochemistry (Formalin/paraffin)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NY-BR-1
NY-BR-1 No.2
Unconjugated
a 77 aa peptide from human NYBR-1 protein
Primary
Purified
Immunohistochemistry
200μg/mL
Mouse
100 μL
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.