missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Milk Fat Globule 1 Human (HMFG1) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Epredia™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

422.00€

Specifications

Antigen Milk Fat Globule 1, Human (HMFG1) Ab-1
Clone EDM45
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12631597
View Documents
Epredia™
MS-512-P1
500 μL Concentrated This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Human milk fat globule membrane protein (HMFG1) is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as on the outer surface of the human milk fat globule. HMFG1 is considered as a differentiation marker. It is useful as specific breast epithelial marker in normal and neoplastic mammary development.

Host Species: Mouse

Clone: EDM45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Delipidated human milk fat globule membrane preparation

Molecular Weight: 40-45kDa

Positive Control: SKBR-3 cells; Breast, colon, or lung tissues

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Milk Fat Globule 1, Human (HMFG1) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Human
IgG2a κ
EDM45
Monoclonal
Mouse
Cancer and Tumor Biology
Delipidated human milk fat globule membrane preparation
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.