missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-13 Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunoprecipitation experiments with Epredia™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody.

151.98€ - 897.60€

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-13
Clone 528 + 199.12
Applications Immunoprecipitation
Classification Monoclonal
Form Concentrated
View More Specs

Products 8
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12681747
View Documents
Epredia™
MS-609-B0
100 μL Biotin 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12691747
View Documents
Epredia™
MS-609-B1
500 μL Biotin 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12661747
View Documents
Epredia™
MS-609-B
1 mL Biotin 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12631757
View Documents
Epredia™
MS-609-P0
100 μL Unconjugated 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12601757
View Documents
Epredia™
MS-609-P1
500 μL Unconjugated 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12621757
View Documents
Epredia™
MS-609-P
1 mL Unconjugated 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
12611757
Afficher les documents
Epredia™
MS-609-P1ABX
100 μL Unconjugated 1mg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
12641757
Afficher les documents
Epredia™
MS-609-PABX
200 μL Unconjugated 1mg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528 + 199.12

Isotype: IgG2a + IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Spécification

Spécification

Epidermal Growth Factor Receptor/EGFR Ab-13
Immunoprecipitation
Concentrated
RUO
Human
IgG2a
528 + 199.12
Monoclonal
Mouse
EGFR Family
Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)
Primary
Vidéos
FDS
Documents

Documents

Product Certifications
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis