Learn More
Epidermal Growth Factor Receptor/EGFR Ab-10 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
Brand: Epredia MS-378-B1
Description
EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.Host Species: Mouse
Clone: 111.6
Isotype: IgG1
Species Reactivity: Human. Not mouse and rat.
Epitope: Extracellular domain
Immunogen: Extracellular domain of human recombinant EGFR protein
Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)
Positive Control: A431 cells; Skin, Placenta or Squamous cell CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native only)
- Effect on EGF Binding (Inhibitory)
Note:
USA: RUO; Int'l: RUO
Specifications
| Epidermal Growth Factor Receptor/EGFR Ab-10 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| EGFR Family | |
| Human | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 111.6 | |
| Biotin | |
| Extracellular domain of human recombinant EGFR protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.