missing translation for 'onlineSavingsMsg'
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-10 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Brand:  Epredia MS-378-B

 View more versions of this product

Product Code. 12651237

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 111.6

Isotype: IgG1

Species Reactivity: Human. Not mouse and rat.

Epitope: Extracellular domain

Immunogen: Extracellular domain of human recombinant EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells; Skin, Placenta or Squamous cell CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)
  • Effect on EGF Binding (Inhibitory)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epidermal Growth Factor Receptor/EGFR Ab-10
Monoclonal
200μg/mL
Mouse
1 mL
EGFR Family
Human
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation
111.6
Biotin
Extracellular domain of human recombinant EGFR protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.