missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 14 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Product Code. 12699146 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
Unit Size:
100µL
1mL
200µL
500µL
7mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Form Concentration unitSize
12699146 500 μL Unconjugated Concentrated 200μg/mL 500µL
12659146 100 μL Biotin Concentrated 200μg/mL 100µL
12669146 500 μL Biotin Concentrated 200μg/mL 500µL
12649146 1 mL Biotin Concentrated 200μg/mL 1mL
12689146 100 μL Unconjugated Concentrated 200μg/mL 100µL
12609156 100 μL Unconjugated Concentrated 1mg/mL 100µL
12629156 200 μL Unconjugated Concentrated 1mg/mL 200µL
12679146 1 mL Unconjugated Concentrated 200μg/mL 1mL
12659156 7 mL Unconjugated Prediluted Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12699146 Supplier Epredia™ Supplier No. MS115P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Mouse

Clone: LL002

Isotype: IgG3

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14 Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LL002
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.