missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL1 Antibody (RAB-C213) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
399.00€ - 849.00€
Specifications
| Antigen | L3MBTL1 |
|---|---|
| Clone | RAB-C213 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611654
|
Novus Biologicals
NBP3-20136-0.025mg |
0.025 mg |
399.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670614
|
Novus Biologicals
NBP3-20136-0.1mg |
0.1 mg |
849.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L3MBTL1 Monoclonal antibody specifically detects L3MBTL1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| L3MBTL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 26013 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| RAB-C213 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry | |
| Unconjugated | |
| Human | |
| Human | |
| dJ138B7.3, FLJ41181, H-L(3)MBT, H-l(3)mbt protein, KIAA0681L3MBTLDKFZp586P1522, l(3)mbt (Drosophila)-like, L(3)mbt protein homolog, L(3)mbt-like, l(3)mbt-like (Drosophila), l(3)mbt-like 1 (Drosophila), L3MBT, lethal (3) malignant brain tumor l(3), lethal(3)malignant brain tumor-like protein | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions. | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title