missing translation for 'onlineSavingsMsg'
Learn More

L3MBTL1 Antibody (RAB-C213), DyLight 594 - Chimeric, Novus Biologicals™

Product Code. 18524759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18524759 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18524759 Supplier Novus Biologicals Supplier No. NBP320137DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

L3MBTL1 Monoclonal antibody specifically detects L3MBTL1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen L3MBTL1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone RAB-C213
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias dJ138B7.3, FLJ41181, H-L(3)MBT, H-l(3)mbt protein, KIAA0681L3MBTLDKFZp586P1522, l(3)mbt (Drosophila)-like, L(3)mbt protein homolog, L(3)mbt-like, l(3)mbt-like (Drosophila), l(3)mbt-like 1 (Drosophila), L3MBT, lethal (3) malignant brain tumor l(3), lethal(3)malignant brain tumor-like protein
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26013
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.