missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1/ORF2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-77929
This item is not returnable.
View return policy
Description
L1/ORF2 Polyclonal specifically detects L1/ORF2 in Human, Chimpanzee samples. It is validated for Western Blot, ELISA.
Specifications
| L1/ORF2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC=2.7.7.49, EC=3.1.21.-, Endonuclease, L1, LINE-1 retrotransposable element ORF2 protein, Long interspersed element-1, ORF2p, Retrovirus-related Pol polyprotein LINE-1, Reverse transcriptase | |
| Chicken | |
| 149.01 kDa | |
| 0.1 mg | |
| Primary | |
| L1/ORF2 Antibody is directed against two regions within the endonuclease domain of L1 ORF2 protein. Reactivity occurs against human L1/ORF2 protein and is useful in determining its presence in various assays. A BLAST analysis was used to suggest cross reactivity with L1/ORF2 proteins from chimpanzee sources based on 100% homology with the immunizing sequences, and from macaque, fruit fly, cattle, dog, opossum, and rat sources based on 69 - 88% homology with the immunizing sequences. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/ml | |
| Western Blot 1:2000-1:10000, ELISA 1:10000 | |
| Q5BJF6 | |
| ODF2 | |
| This IgY fraction L1/ORF2 Antibody was prepared from eggs of chickens laid after repeated immunizations with two synthetic peptides conjugated to keyhole limpet hemocyanin (KLH). The peptides correspond to regions within the endonuclease domain of L1/ORF2 protein. (Uniprot: O00370) | |
| Affinity Purified | |
| RUO | |
| 4957 | |
| Human, Chimpanzee | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction