missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1/ORF2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-44223
This item is not returnable.
View return policy
Description
Specifically detects L1/ORF2 in Mouse samples, and it is validated for ELISA
L1/ORF2 Polyclonal specifically detects L1/ORF2 in Mouse samples. It is validated for ELISA.
Specifications
| L1/ORF2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC=2.7.7.49, EC=3.1.21.-, Endonuclease, L1, LINE-1 retrotransposable element ORF2 protein, Long interspersed element-1, ORF2p, Retrovirus-related Pol polyprotein LINE-1, Reverse transcriptase | |
| This IgY fraction L1/ORF2 Antibody was prepared from eggs of chickens laid after repeated immunizations with three synthetic peptides conjugated to keyhole limpet hemocyanin (KLH). The peptides correspond to regions within the endonuclease domain of L1/ORF2 protein. (Uniprot: P11369) | |
| Multi-step | |
| RUO | |
| Anti-L1/ORF2 Antibody is directed against three regions within the endonuclease domain of L1 ORF2 protein. Reactivity occurs against mouse L1/ORF2 protein and is useful in determining its presence in various assays. A BLAST analysis shows reactivity with L1/ORF2 proteins from mouse sources based on 100% homology with the immunizing sequences. Cross reactivity against L1/ORF2 from other sources is unknown. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 1.21 mg/mL | |
| ELISA 1:20000-1:100000 | |
| P11369 | |
| Chicken | |
| 149.58 kDa | |
| 0.1 mg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction