missing translation for 'onlineSavingsMsg'
Learn More

L-idose (0.145M solution), TRC

Product Code. 30179576
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179576 5 mg 5mg
30179536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179576 Supplier Toronto Research Chemicals Supplier No. TRCI1750605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-56-5
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym L-Idose, Idose, L- (8CI)
IUPAC Name (2R,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5+,6+/m0/s1
Formula Weight 180.0633
Concentration or Composition (by Analyte or Components) L-Idose 0.145M
Chemical Name or Material L-Idose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.