CAS RN 5934-56-5
CAS RN 5934-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-idose (0.145M solution), TRC
CAS: 5934-56-5 Molecular Formula: C6 H12 O6 Molecular Weight (g/mol): 180.16 Synonym: L-Idose,Idose, L- (8CI) IUPAC Name: (2R,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal SMILES: OC[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O