missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KvBeta2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | KvBeta2 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
KvBeta2 Polyclonal specifically detects KvBeta2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| KvBeta2 | |
| Unconjugated | |
| RUO | |
| NP_742128 | |
| 8514 | |
| Synthetic peptide directed towards the middle region of human KCNAB2. Peptide sequence: SSVLLGASNADQLMENIGAIQVLPKLSSSIIHEIDSILGNKPYSKKDYRS | |
| Primary |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| HKvbeta2, HKvbeta2.1, HKvbeta2.2, K(+) channel subunit beta-2, K+ channel beta-2 subunit, KCNA2BAKR6A5, KCNK2, KV-BETA-2, MGC117289, potassium channel shaker chain beta 2, potassium voltage-gated channel, shaker-related subfamily, beta member 2, voltage-gated potassium channel subunit beta-2 | |
| KCNAB2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title