Learn More
Kv4.2 Antibody (5B11B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52497
This item is not returnable.
View return policy
Description
Kv4.2 Monoclonal specifically detects Kv4.2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Kv4.2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
| Mouse | |
| 70.5 kDa | |
| 0.1 mg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3751 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| 5B11B9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| KCND2 | |
| Purified recombinant fragment of human KCND2 (AA: 27-184) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |