missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KU70/XRCC6 Mouse anti-Human, Mouse, Rat, Clone: 3C12D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66607-1-IG-20UL
This item is not returnable.
View return policy
Description
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.Specifications
| KU70/XRCC6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12956, P23475 | |
| Xrcc6 | |
| KU70,XRCC6 Fusion Protein Ag21301 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3C12D9 | |
| Unconjugated | |
| XRCC6 | |
| 5 dRP lyase Ku70, 70 kDa subunit of Ku antigen, CTC75, CTCBF, DNA repair protein XRCC6, G22P1, KU70, ML8, Thyroid lupus autoantigen, TLAA, XRCC6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 14375, 25019, 2547 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction