missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSHV ORF26 Antibody (2F6B8), Novus Biologicals™
Mouse Monoclonal Antibody
161.00€
Specifications
| Antigen | KSHV ORF26 |
|---|---|
| Clone | 2F6B8 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
KSHV ORF26 Monoclonal antibody specifically detects KSHV ORF26 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| KSHV ORF26 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| HHV8, Human Herpes Virus 8, Kaposi's sarcoma herpes virus, KSHV, ORF45 | |
| ORF26 | |
| IgG1 | |
| Unpurified |
| 2F6B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| 4961509 | |
| Purified recombinant fragment of KSHV ORF26 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title