missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTAP2-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 589.00€
Specifications
| Antigen | KRTAP2-4 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245042
|
Novus Biologicals
NBP2-57069 |
100 μL |
589.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681819
|
Novus Biologicals
NBP2-57069-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KRTAP2-4 Polyclonal specifically detects KRTAP2-4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| KRTAP2-4 | |
| Polyclonal | |
| Rabbit | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 85294 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PVTCQTTVCRPVTCVPRCTRPICEPC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| High sulfur keratin-associated protein 2.4, KAP2.4MGC119884, keratin associated protein 2-4, Keratin-associated protein 2.4, keratin-associated protein 2-4, KRTAP2.4, MGC119882, MGC119885, MGC74790 | |
| KRTAP2-4 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title