missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT7 Monoclonal Antibody (OTI3E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803333
This item is not returnable.
View return policy
Description
Blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).Specifications
| KRT7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08729 | |
| KRT7 | |
| Human recombit protein fragment corresponding to amino acids 1-114 and 340-469 of human KRT7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E5 | |
| Unconjugated | |
| KRT7 | |
| CK7, K2C7, K7, SCL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3855 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction