missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KREMEN1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148569
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
KREMEN1 is a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. This protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains.
Specifications
| KREMEN1 | |
| Polyclonal | |
| Unconjugated | |
| Kremen1 | |
| AV002070; dickkopf receptor; KREMEN; kremen protein 1; KREMEN1; kringle containing transmembrane protein 1; kringle domain-containing transmembrane protein 1; kringle-coding gene marking the eye and the nose; kringle-containing protein marking the eye and the nose; KRM1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 83999 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96MU8 | |
| Kremen1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kremen-1, Ala20-Gly397. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction