missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNB1 Monoclonal Antibody (OTI9D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811714
This item is not returnable.
View return policy
Description
Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.Specifications
| KPNB1 | |
| Monoclonal | |
| Unconjugated | |
| KPNB1 | |
| IMB1, IPO1, IPOB, Impnb, NTF97 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 16211, 24917, 3837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI9D6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52296, P70168, Q14974 | |
| Kpnb1 | |
| Human recombit protein fragment corresponding to amino acids 1-200 of human KPNB1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction