missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Antibody (CL5346), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61416-25ul
This item is not returnable.
View return policy
Description
KMT2D Monoclonal specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| KMT2D | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| MLL2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AQLADTLFSKGLGPWDPPDNLAETQKPEQSSLVPGHLDQVNGQVVPEASQLSIKQEPREEPCALGAQSVKREANGEPIGAPGTSNHLLLAGPRSEAGHLLLQKLLRAKNVQLSTGRGSEGLRAEINGHI | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence | |
| CL5346 | |
| Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8085 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction