missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRA1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
575.00€
Specifications
| Antigen | KLRA1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Form | Purified |
Description
KLRA1 Polyclonal antibody specifically detects KLRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| KLRA1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of human KLRA1. The immunogen is located within amino acids 190 - 240 of KLRA1. Amino Acid Squence: IGLSYDERESKWK | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 500 SE | |
| Rabbit | |
| Immunology | |
| 50mM Sodium Borate | |
| 10748 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title