missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL15 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€
Specifications
| Antigen | KLHL15 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
KLHL15 Polyclonal specifically detects KLHL15 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| KLHL15 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| NP_085127 | |
| 80311 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human KLHL15. The immunogen is located within the last 50 amino acids of KLHL15. Amino Acid Squence: DPDENKKWLEDEYPRM | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| kelch-like 15 (Drosophila), kelch-like protein 15, KIAA1677, MGC126148, MGC126149 | |
| KLHL15 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title