missing translation for 'onlineSavingsMsg'
Learn More

KIR3DL3/CD158z Antibody (1136B) [Biotin], Novus Biologicals™

Product Code. 18661605 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18661605 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18661605 Supplier Novus Biologicals Supplier No. FAB8919B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DL3/CD158z
Applications Flow Cytometry
Classification Monoclonal
Clone 1136B
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z, Met1-Leu322, Accession # Q8N743
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115653
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.