missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen 1 Antibody (KNG1/7422), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44613-20ug
This item is not returnable.
View return policy
Description
Kininogen 1 Monoclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for Immunohistochemistry
Especificaciones
| Kininogen 1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| KNG1/7422 | |
| Unconjugated | |
| Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
| Recombinant fragment (around aa1-200) of human Kininogen 1 protein (Uniprot: P01042). Exact sequence is proprietary. | |
| 20 μg | |
| Apoptosis, Immunology | |
| 3827 | |
| Store at 4⪚C. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido