missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5A Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32231B
This item is not returnable.
View return policy
Description
Kinesin 5A Polyclonal antibody specifically detects Kinesin 5A in Human samples. It is validated for Western Blot
Specifications
| Kinesin 5A | |
| Polyclonal | |
| Western Blot | |
| D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10 | |
| The immunogen this antibody was made to, maps to a region between residue 982 to 1032 of human Kinesin Family Member 5A using the numbering given in entry NP_004975.2 (GeneID 3798). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3798 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction