Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ KimCote™ Heavy-Duty Serum Bottles
Product Code. p-3827191
Change view
Click to view available options
Capacity (Metric):
4000 mL
9000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11797969 4000 mL Each
11727979 9000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11797969 Supplier DWK Life Sciences Supplier No. KC149604
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Plastic-coated for added safety
  • KimCote™ plastic coating for added safety in the event of breakage
  • Autoclavable
  • Ultra-clear and extremely durable
  • Resistant to many common laboratory chemicals
  • Unique texture provides non-slip handling, wet or dry
TRUSTED_SUSTAINABILITY
Specifications
Capacity 135.2 oz., 4000mL
Capacity (Metric) 4000 mL
Capacity (English) 135.2 oz.
Color Clear
Diameter (English) 6.57 in.
Material Glass
Height (Metric) 310 mm
Height (English) 12.2 in.
Closure Material Rubber
Closure Size No. 8
Diameter (Metric) 167 mm