Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Product Code. 11772404
Click to view available options
Diameter (Metric) Inner:
1 cm
1.5 cm
2.5 cm
Unit Size:
Each
Product Code. 11772404
Brand: DWK Life Sciences 4204042500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
  • A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
  • A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 2.5 cm
For Use With Kimble-Chase Economy Columns