missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody (SR00-02), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32498
This item is not returnable.
View return policy
Description
Ki67/MKI67 Monoclonal antibody specifically detects Ki67/MKI67 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Ki67/MKI67 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:5000-1:150000 | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| Synthetic peptide within human Ki67/MKI67 aa 1,040-1,080. (Uniprot: P46013) | |
| 100 μL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| 4288 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| SR00-02 | |
| Unconjugated | |
| PBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction