missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketorolac EP Impurity H, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ketorolac EP Impurity H
Specifications
Specifications
| Chemical Name or Material | Ketorolac Methyl Ester |
| CAS | 80965-09-9 |
| Analyte or Component Names | Ketorolac Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl (1RS)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (Ketorolac Methyl Ester), Ketorolac Trometamol Imp. H (EP), 5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid methyl ester, Ketorolac methyl ester, Ketorolac Trometamol Impurity H, Ketorolac Impurity H |
| InChI Formula | InChI=1S/C16H15NO3/c1-20-16(19)12-9-10-17-13(12)7-8-14(17)15(18)11-5-3-2-4-6-11/h2-8,12H,9-10H2,1H3 |
| SMILES | COC(=O)C1CCn2c1ccc2C(=O)c3ccccc3 |
| IUPAC Name | methyl 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction