missing translation for 'onlineSavingsMsg'
Learn More

Kerosene Blank, VHG™

Product Code. 17990456
Change view
Click to view available options
Quantity:
500 mL
1 gal
Unit Size:
1 GAL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990456 1 gal 1 GAL
17922356 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17990456 Supplier VHG Labs Supplier No. VHGKERO1GAL
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Kerosene Blank
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kerosene Blank
Methods ICP/XRF/RDE
Analyte or Component Names Kerosene Blank
Shipping Condition Room Temperature
IUPAC Name Kerosene Blank
Solvent or Matrix Kerosene
Quantity 1 gal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.