missing translation for 'onlineSavingsMsg'
Learn More

Kerosene Blank, VHG™

Product Code. 17922356
Change view
Click to view available options
Quantity:
500 mL
1 gal
Unit Size:
1 GAL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922356 500 mL 500mL
17990456 1 gal 1 GAL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17922356 Supplier VHG Labs Supplier No. VHGKERO500

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Kerosene Blank
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kerosene Blank
Methods ICP/XRF/RDE
Analyte or Component Names Kerosene Blank
Shipping Condition Room Temperature
IUPAC Name Kerosene Blank
Solvent or Matrix Kerosene
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.