missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 493.00€
Specifications
| Antigen | KDM6A |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18349492
|
Novus Biologicals
NBP3-05103-20UL |
20 μL |
216.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18379051
|
Novus Biologicals
NBP3-05103-100UL |
100 μL |
493.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KDM6A Polyclonal specifically detects KDM6A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| KDM6A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 7403 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| bA386N14.2, bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeatprotein (UTX)), DKFZp686A03225, EC 1.14.11, EC 1.14.11.-, Histone demethylase UTX, lysine (K)-specific demethylase 6A, MGC141941, ubiquitously transcribed tetratricopeptide repeat protein X-linked, ubiquitously transcribed tetratricopeptide repeat, X chromosome, ubiquitously transcribed TPR protein on the X chromosome, ubiquitously transcribed X chromosome tetratricopeptide repeat protein, ubiquitously-transcribed TPR gene on the X chromosome, Ubiquitously-transcribed TPR protein on the X chromosome, Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein, UTXlysine-specific demethylase 6A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 550-700 of mouse KDM6A (NP_033509.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title