missing translation for 'onlineSavingsMsg'
Learn More

Abcam KDM6A/KDM6B Activity Quantification Assay Kit, Fluorometric, 48 Tests

Product Code. 32109650
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
48 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109650 48 Tests 48 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109650 Supplier Abcam Supplier No. ab15691148Test
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Measure KDM6A and KDM6B activity in cell or tissue extracts within 3 hours using a fluorometric detection method.

  • Assay Type: Enzyme activity
  • Detection Method: Fluorescent
  • Assay Time: 3 hr
  • Sensitivity: 10 ng
  • Sample Types: Nuclear Extracts, Purified protein
  • Applications: Enzyme activity assay

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Western Blot Kit
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 48 Tests
Run Time 3 hr.
Target Molecule KDM6A, KDM6B
Detection Method Fluorescence
Sample Type Nuclear Extracts, Purified protein
Quantity 48 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.