Learn More
KCTD21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81563
This item is not returnable.
View return policy
Description
KCTD21 Polyclonal specifically detects KCTD21 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| BTB/POZ domain-containing protein KCTD21, KCASH2, potassium channel tetramerisation domain containing 21 | |
| IgG |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |