Learn More
KCTD21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-81563
 View more versions of this product
Product Code. 18768193
  • 624.00€ / 0.10mL
Description
KCTD21 Polyclonal specifically detects KCTD21 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
BTB/POZ domain-containing protein KCTD21, KCASH2, potassium channel tetramerisation domain containing 21
IgG
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.